Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.995% (metals basis)

$400.22 - $400.22

Chemical Identifiers

Chemical Name or Material Tin sputtering target
CAS 7440-31-5
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00133862
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40899KS

 
alfa aesar™
40899KS
1 each Each for $400.22
Description & Specifications

Specifications

Chemical Name or Material Tin sputtering target
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay 99.995%
Assay Percent Notes (metals basis)
MDL Number MFCD00133862
CAS 7440-31-5
Odor Odorless
Quantity 1 each