Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.995% (metals basis)

Magnetron sputtering source

$194.38 - $194.38

Chemical Identifiers

Chemical Name or Material Tin sputtering target
CAS 7440-31-5
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00133862
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40898KS

  SDS
alfa aesar™
40898KS
1 each Each for $194.38
Description & Specifications

Specifications

Chemical Name or Material Tin sputtering target
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.995%
Assay Percent Notes (metals basis)
MDL Number MFCD00133862
CAS 7440-31-5
Odor Odorless
Quantity 1 each