missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin slug, 11mm (0.43 in.) dia. x 4mm (0.2 in.) thick, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 14134KD

 View more versions of this product

Catalog No. AA14134KD



Specifications

Specifications

Tin slug
50 pc.
7440-31-5
Sn
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
[Sn]
Odorless
5352426
Slug
Puratronic™
≈2.6g
11mm (0.43 in.) dia. x 4mm (0.2 in.) thick
(metals basis)
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
118.71
CHEBI:32990
99.999%

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
CHEBI:32990
[Sn]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents