missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin shot, 3mm (0.1 in.), Puratronic™, 99.9999% (metals basis)

Manufacturer:  Alfa Aesar™ 1100922

 View more versions of this product

Catalog No. AA1100922



Specifications

Specifications

Tin shot
3mm (0.1 in.)
(metals basis)
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
118.71
CHEBI:32990
99.9999%
100g
7440-31-5
Sn
flake
[Sn]
Odorless
5352426
Teardrop Shot
Puratronic™

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
flake
CHEBI:32990
[Sn]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents