Learn More
Tin selenide, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01878106
Description
Tin selenide finds application in low- cost photovoltaics devices and memory-switching devices. It acts as a solid-state lubricant. It is also used for solar cells and anodes for lithium-ion batteries. Further, it is a narrow band-gap semiconductor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tin selenide | |
1315-06-6 | |
5 g | |
SnSe | |
Insoluble in water. Soluble in aqua regia,alkali sulfides,and alkali selenides. |
861°C | |
Pieces | |
99.999% (metals basis) | |
MFCD00054049 | |
197.65 |
Chemical Identifiers
1315-06-6 | |
SnSe | |
MFCD00054049 |
Safety and Handling
P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
H301+H331-H373
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 215-257-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only