Please login to your online account to display your discounted pricing

Tin, reagent ACS, shot, <100 mesh, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC421001000

 View more versions of this product

Catalog No. AC421001000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Shot
Assay ≥99.5%
Purity Grade ACS Reagent Grade
Formula Weight 118.69
Quantity 100g
Color Silvery-gray
EINECS Number 231-141-8
TSCA TSCA
Density 7.3000g/mL
RTECS Number XP7320000
Fieser 01,1168; 02,414; 11,519; 12,486; 13,298; 17,333
Merck Index 15, 9603
Iron (Fe) 100ppm max.
Packaging Glass bottle
Arsenic (As) 1ppm max.
Chemical Name or Material Tin
Name Note <100 mesh
MDL Number MFCD00133862
Antimony (Sb) 200ppm max.
Mesh Size 8 to 20 mesh
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Copper (Cu) 50ppm max.
Molecular Formula Sn
Lead (Pb) 50ppm max.
Linear Formula Sn
CAS 7440-31-5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tin
Name Note <100 mesh
CAS 7440-31-5
Purity Grade ACS Reagent Grade
Physical Form Shot
Assay ≥99.5%
Molecular Formula Sn
Linear Formula Sn
Formula Weight 118.69
MDL Number MFCD00133862
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.

SDS & Safety Documents