Please login to your online account to display your discounted pricing

Tin, reagent ACS, shot, <100 mesh, ACROS Organics™

$90.58 - $90.58

Chemical Identifiers

Chemical Name or Material Tin
Name Note <100 mesh
CAS 7440-31-5
Purity Grade ACS Reagent Grade
Physical Form Shot
Assay ≥99.5%
Molecular Formula Sn
Linear Formula Sn
Formula Weight 118.69
MDL Number MFCD00133862
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC421001000

  SDS
thermo fisher scientific
AC421001000
Glass bottle 100g Each for $90.58

AC421005000

  SDS
fisher scientific acros
421005000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Shot
Assay ≥99.5%
Formula Weight 118.69
Purity Grade ACS Reagent Grade
Quantity 100g
Color Silvery-gray
TSCA TSCA
RTECS Number XP7320000
EINECS Number 231-141-8
Density 7.3000g/mL
Fieser 01,1168; 02,414; 11,519; 12,486; 13,298; 17,333
Merck Index 15, 9603
Iron (Fe) 100ppm max.
Packaging Glass bottle
Arsenic (As) 1ppm max.
Mesh Size 8 to 20 mesh
Chemical Name or Material Tin
MDL Number MFCD00133862
Name Note <100 mesh
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Antimony (Sb) 200ppm max.
Lead (Pb) 50ppm max.
Copper (Cu) 50ppm max.
Linear Formula Sn
CAS 7440-31-5
Molecular Formula Sn
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Reduction reagent

Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.