Please login to your online account to display your discounted pricing

Tin, reagent ACS, shot, <100 mesh, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: fisher scientific acros  421005000

 View more versions of this product

Catalog No. AC421005000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.5%
Purity Grade ACS Reagent Grade
Physical Form Shot
Formula Weight 118.69
Quantity 500g
Color Silvery-gray
Merck Index 15, 9603
EINECS Number 231-141-8
TSCA TSCA
Density 7.3000g/mL
RTECS Number XP7320000
Fieser 01,1168; 02,414; 11,519; 12,486; 13,298; 17,333
Copper (Cu) 50ppm max.
Linear Formula Sn
MDL Number MFCD00133862
CAS 7440-31-5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Tin
Name Note <100 mesh
Iron (Fe) 100ppm max.
Lead (Pb) 50ppm max.
Antimony (Sb) 200ppm max.
Mesh Size 8 to 20 mesh
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Arsenic (As) 1ppm max.
Molecular Formula Sn

Specifications

Chemical Name or Material Tin
Name Note <100 mesh
CAS 7440-31-5
Purity Grade ACS Reagent Grade
Physical Form Shot
Assay ≥99.5%
Molecular Formula Sn
Linear Formula Sn
Formula Weight 118.69
MDL Number MFCD00133862
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.

SDS & Safety Documents