Please login to your online account to display your discounted pricing

Tin protoporphyrin IX dichloride, Tocris

Heme oxygenase inhibitor

$149.90 - $632.23

Chemical Identifiers

Chemical Name or Material Tin protoporphyrin IX dichloride
Assay Percent Range >98%
Molecular Formula C34H32Cl2N4O4Sn
Formula Weight 750.25
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

074750

 
r & d systems
074750
50mg Each for $632.23

074710

 
r & d systems
074710
10mg Each for $149.90
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 750.25
Chemical Name or Material Tin protoporphyrin IX dichloride
Molecular Formula C34H32Cl2N4O4Sn
CAS 14325-05-4
Assay Percent Range >98%
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature