Please login to your online account to display your discounted pricing

Tin protoporphyrin IX dichloride, Tocris


Heme oxygenase inhibitor

Quantity 50mg

Manufacturer: r & d systems  074750

 View more versions of this product

Catalog No. 074750

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 750.25
CAS 14325-05-4
Chemical Name or Material Tin protoporphyrin IX dichloride
Molecular Formula C34H32Cl2N4O4Sn
Assay Percent Range >98%
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Tin protoporphyrin IX dichloride
Assay Percent Range >98%
Molecular Formula C34H32Cl2N4O4Sn
Formula Weight 750.25
Accessories

Store at Room Temperature