missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin Powder, -325 mesh, 99.8% (Metals basis), Alfa Aesar™

Apparent density 3.2g/cm3

Manufacturer:  Thermo Fisher Scientific 1037922

 View more versions of this product

Catalog No. AA1037922



Specifications

Specifications

Tin
7440-31-5
99.8% (Metals basis)
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
118.71
CHEBI:32990
Gray
100g
Gray powder
Sn
flake
[Sn]
Odorless
5352426
Powder

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
flake
CHEBI:32990
[Sn]
Safety and Handling

Safety and Handling

H228; Flammable solid.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-141-8

Documents