missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin Powder, -325 mesh, 99.8% (Metals basis), Alfa Aesar™

Apparent density 3.2g/cm3

Manufacturer:  Thermo Fisher Scientific 10379A3

 View more versions of this product

Catalog No. AA10379A3



Specifications

Specifications

Tin
7440-31-5
99.8% (Metals basis)
MFCD00133862
flake, metallic, powder, silver matt powder, stannum, tin, elemental, tin, metal, wang, zinn, zinn german
[Sn]
Odorless
5352426
Powder, Apparent density 3.2g/cm
2kg
Gray powder
Sn
-325 Mesh
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
118.71
CHEBI:32990
Gray

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
flake, metallic, powder, silver matt powder, stannum, tin, elemental, tin, metal, wang, zinn, zinn german
CHEBI:32990
[Sn]
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P314-P330-P501c

H302-H319-H335-H336-H373-H500

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-141-8

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents