missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin powder, -100 mesh, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 1037822

 View more versions of this product

Catalog No. AA1037822



Specifications

Specifications

Tin powder
7440-31-5
Sn
−100 Mesh
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
118.71
CHEBI:32990
99.5%
100g
(metals basis)
MFCD00133862
flake
[Sn]
Odorless
5352426
Powder

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
flake
CHEBI:32990
[Sn]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents