Alfa Aesar™ Tin plating solution, electroless, part B
Manufacturer:  Alfa Aesar™ 441772Y
 View more versions of this product
Catalog No. AA441772Y
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • To preserve solderability of printed circuit boards, for plating electronic parts, and for corrosion protection
Specifications
Specifications
Tin plating solution
Electroless, part B
UN1789
Liquid
4x1pt.
MFCD03703331
Acrid
Colorless
Documents