missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin plate 3.2mm (0.13 in.) thick, 99.85% (metals basis)

Manufacturer:  Alfa Aesar™ 12602GH

 View more versions of this product

Catalog No. AA12602GH



Specifications

Specifications

Tin plate
100 x 100mm
7440-31-5
Sn
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
[Sn]
Odorless
5352426
Plate
≈234g/100x100mm
3.2mm (0.13 in.) thick
(metals basis)
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
118.71
CHEBI:32990
99.85%

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00133862
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
CHEBI:32990
[Sn]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents