Learn More
Alfa Aesar™ Tin, plasma standard solution, Specpure™, Sn 10,000μg/mL
Manufacturer: Alfa Aesar™ 14384AD
Specifications
Tin plasma standard solution | |
MFCD00133862 | |
Sn in 20% HCl | |
Specpure™ |
Sn 10,000μg/mL | |
UN1789 | |
Liquid | |
50mL |
Chemical Identifiers
MFCD00133862 |
Safety and Handling
GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID
TSCA : TSCA