Learn More
Tin, plasma standard solution, Specpure™ Sn 10,000μg/mL, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 014384AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tin plasma standard solution | |
Sn 10,000μg/mL | |
MFCD00133862 | |
Specpure™ |
Sn in 20% HCl | |
100 mL | |
UN1789 | |
Liquid |
Chemical Identifiers
MFCD00133862 |
Safety and Handling
GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID
EINECSNumber : 231-595-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only