Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin, Oil based standard solution, Specpure™, Sn 1000μg/g

Chemical Identifiers

Chemical Name or Material Tin Oil based standard solution
Name Note Sn 1000μg/g
Purity Grade Specpure™
MDL Number MFCD00133862
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1420618

  SDS
alfa aesar™
1420618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

MDL Number MFCD00133862
Quantity 50g
Chemical Name or Material Tin Oil based standard solution
Physical Form Liquid
Name Note Sn 1000μg/g
Purity Grade Specpure™