Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin(IV) tert-pentyloxide, 5% w/v in 2-methyl-2-butanol, 99% (metals basis)

Chemical Identifiers

Chemical Name or Material Tin(IV) tert-pentyloxide
Formula Weight 467.27
MDL Number MFCD00145409
UN Number UN3274
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA36562AA

  SDS
alfa aesar™
36562AA
5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA36562AC

  SDS
alfa aesar™
36562AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA36562AE

  SDS
alfa aesar™
36562AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 467.27
Chemical Name or Material Tin(IV) tert-pentyloxide
Boiling Point 100°C (0.2mmHg)
Physical Form Liquid
Assay 99%
Assay Percent Notes (metals basis)
MDL Number MFCD00145409
Quantity 5mL
UN Number UN3274
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H314-H318-H302-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Concentration 5% w/v in 2-methyl-2-butanol
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.
Sensitivity Moisture Sensitive
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H318-H302-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.