Learn More
Tin(IV) phthalocyanine dichloride, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03960603
Description
Tin(IV) phthalocyanine dichloride is a stable metal phthalocyanine dye utilized in biochemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tin(IV) phthalocyanine dichloride | |
C32H16Cl2N8Sn | |
1 g | |
Moisture sensitive | |
Insoluble in water. | |
C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C6N4[Sn](N7C(=NC2=N3)C8=CC=CC=C8C7=NC9=NC(=N6)C1=CC=CC=C19)(Cl)Cl | |
9580361 | |
Powder |
18253-54-8 | |
MFCD00054210 | |
UN3146 | |
tin iv phthalocyanine dichloride, sncl2pc, dichlorotin phthalocyanine, tin phthalocyanine dichloride, tin iv phthalocyaninedichloride | |
BBRNKSXHHJRNHK-UHFFFAOYSA-L | |
702.146 | |
702.34 |
Chemical Identifiers
18253-54-8 | |
702.146 | |
BBRNKSXHHJRNHK-UHFFFAOYSA-L | |
9580361 |
C32H16Cl2N8Sn | |
MFCD00054210 | |
tin iv phthalocyanine dichloride, sncl2pc, dichlorotin phthalocyanine, tin phthalocyanine dichloride, tin iv phthalocyaninedichloride | |
C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C6N4[Sn](N7C(=NC2=N3)C8=CC=CC=C8C7=NC9=NC(=N6)C1=CC=CC=C19)(Cl)Cl |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only