missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin(IV) oxide, nanopowder

Manufacturer:  Alfa Aesar™ 4460609

 View more versions of this product

Catalog No. AA4460609



Specifications

Specifications

Tin (IV) oxide
O2Sn
MFCD00011244
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
O=[Sn]=O
150.708
CHEBI:52991
Nanopowder
Odorless
1.9968
1630°C
18282-10-5
SnO2
14,8778
XOLBLPGZBRYERU-UHFFFAOYSA-N
dioxotin
29011
150.69
6.95g/mL
8 to 10nm APS
1800°C to 1900°C (sublimation)
10g

Chemical Identifiers

18282-10-5
150.708
XOLBLPGZBRYERU-UHFFFAOYSA-N
29011
dioxotin
O2Sn
MFCD00011244
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
CHEBI:52991
O=[Sn]=O
Safety and Handling

Safety and Handling

RTECSNumber : XQ4000000

TSCA : TSCA

SDS Documents