missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin(IV) oxide, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1228322

 View more versions of this product

Catalog No. AA1228322



Specifications

Specifications

18282-10-5
O2Sn
cassiterite
XOLBLPGZBRYERU-UHFFFAOYSA-N
dioxotin
29011
150.69
6.95
1.9968
White to Gray
100g
99.9% metal basis
MFCD00011244
Soluble in hot concentrated alkalis,and concentrated acids. Insoluble in alcohol.
O=[Sn]=O
150.708
CHEBI:52991
<10 Micron Powder
Odorless
1800°C to 1900°C (sublimation)
1630°C

Chemical Identifiers

18282-10-5
150.708
XOLBLPGZBRYERU-UHFFFAOYSA-N
29011
dioxotin
O2Sn
MFCD00011244
cassiterite
CHEBI:52991
O=[Sn]=O
Safety and Handling

Safety and Handling

EINECSNumber : 242-159-0

RTECSNumber : XQ4000000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents