missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin(IV) oxide, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1228336

 View more versions of this product

Catalog No. AA1228336



Specifications

Specifications

18282-10-5
O2Sn
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
O=[Sn]=O
150.708
CHEBI:52991
Powder
White to Gray
99.9% metal basis
MFCD00011244
XOLBLPGZBRYERU-UHFFFAOYSA-N
dioxotin
29011
150.69
Odorless
500g

Chemical Identifiers

18282-10-5
150.708
XOLBLPGZBRYERU-UHFFFAOYSA-N
29011
dioxotin
O2Sn
MFCD00011244
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
CHEBI:52991
O=[Sn]=O
Documents