missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin(IV) isopropoxide, 99% (metals basis), 10% w/v in isopropanol

Manufacturer:  Alfa Aesar™ 45480AE

 View more versions of this product

Catalog No. AA45480AE



Specifications

Specifications

Tin (IV) isopropoxide
100mL
10% w/v in isopropanol
Moisture Sensitive
C12H32O4Sn
MFCD00145408
tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate
CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn]
359.094
355.06
99%
1184-61-8
Alcohol-like
Colorless
(metals basis)
Sn[OCH(CH3)2]4
UN3274
VUDAJANAMZGVOP-UHFFFAOYSA-N
propan-2-ol;tin
85354423
Liquid

Chemical Identifiers

1184-61-8
359.094
VUDAJANAMZGVOP-UHFFFAOYSA-N
85354423
CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn]
C12H32O4Sn
MFCD00145408
tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate
propan-2-ol;tin
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H302-H336
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.

SDS Documents