Learn More
Tin(IV) isopropoxide, 99% (metals basis), 10% w/v in isopropanol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 045480AC
Description
Tin(IV) isopropoxide is used as a precursor for the preparation of tin oxide nanoparticles, thin films, indium tin oxide (ITO) films , which shows desired electrochemical and conducting properties. Doped tin oxide nanoparticles and thin films find application as gas sensors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tin (IV) isopropoxide | |
Colorless | |
(metals basis) | |
Sn[OCH(CH3)2]4 | |
25 mL | |
Moisture sensitive | |
Miscible with alcohols,ketones and esters. | |
CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn] | |
359.094 | |
355.06 | |
Liquid |
1184-61-8 | |
Alcohol-like | |
C12H32O4Sn | |
MFCD00145408 | |
UN3274 | |
tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate | |
VUDAJANAMZGVOP-UHFFFAOYSA-N | |
propan-2-ol;tin | |
85354423 | |
99% |
Chemical Identifiers
1184-61-8 | |
359.094 | |
VUDAJANAMZGVOP-UHFFFAOYSA-N | |
85354423 | |
CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn] |
C12H32O4Sn | |
MFCD00145408 | |
tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate | |
propan-2-ol;tin |
Safety and Handling
GHS H Statement
H225-H314-H302-H336
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only