missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) isopropoxide, 98% (metals basis), 10% w/v in isopropanol/toluene

CAS: 1184-61-8 | C12H32O4Sn | 359.094 g/mol

Supplier:  Thermo Scientific Chemicals 036563AC

 View more versions of this product

Catalog No. AA36563AC


Only null left
Explore available promotions

Description

Description

Tin(IV) isopropoxide isopropanol is used has a important tin precursor for the synthesis of tin oxide nanoparticles. Used in Indium Tin Oxide films, thin films doped tin oxide nanoparticles, thin films and are widely studied as gas sensors. Tin(IV) precursor solution was prepared from tin(IV) isopropoxide dissolved in isopropyl alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tin(IV) isopropoxide
(metals basis)
MFCD00145408
UN1993
tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate
VUDAJANAMZGVOP-UHFFFAOYSA-N
propan-2-ol;tin
85354423
98%
1184-61-8
C12H32O4Sn
25 mL
Moisture sensitive
Soluble in hydrocarbons,reacts with alcohols,ketones and esters
CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn]
359.094
355.06
Liquid

Chemical Identifiers

1184-61-8
359.094
VUDAJANAMZGVOP-UHFFFAOYSA-N
85354423
CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn]
C12H32O4Sn
MFCD00145408
tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate
propan-2-ol;tin
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H304-H314-H335-H336-H361d-H373

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only