Learn More
Tin(IV) isopropoxide, 98% (metals basis), 10% w/v in isopropanol/toluene, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 036563AP
Description
Tin(IV) isopropoxide isopropanol is used has a important tin precursor for the synthesis of tin oxide nanoparticles. Used in Indium Tin Oxide films, thin films doped tin oxide nanoparticles, thin films and are widely studied as gas sensors. Tin(IV) precursor solution was prepared from tin(IV) isopropoxide dissolved in isopropyl alcohol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tin(IV) isopropoxide | |
(metals basis) | |
MFCD00145408 | |
UN1993 | |
tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate | |
VUDAJANAMZGVOP-UHFFFAOYSA-N | |
propan-2-ol;tin | |
85354423 | |
98% |
1184-61-8 | |
C12H32O4Sn | |
500 mL | |
Moisture sensitive | |
Soluble in hydrocarbons,reacts with alcohols,ketones and esters | |
CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn] | |
359.094 | |
355.06 | |
Liquid |
Chemical Identifiers
1184-61-8 | |
359.094 | |
VUDAJANAMZGVOP-UHFFFAOYSA-N | |
85354423 | |
CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn] |
C12H32O4Sn | |
MFCD00145408 | |
tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate | |
propan-2-ol;tin |
Safety and Handling
GHS H Statement
H225-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H304-H314-H335-H336-H361d-H373
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only