Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin(IV) iodide, ultra dry, 99.998% (metals basis)

Chemical Identifiers

Chemical Name or Material Tin(IV) iodide
CAS 7790-47-8
Name Note Ultra dry
Formula Weight 626.31
MDL Number MFCD00036272
UN Number UN3260
Merck Index 14,8777
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1361706

  SDS
alfa aesar™
1361706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1361714

  SDS
alfa aesar™
1361714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Tin(IV) iodide
Name Note Ultra dry
CAS 7790-47-8
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Ampouled under argon
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
TSCA TSCA
Physical Form Powder
Assay 99.998%
Assay Percent Notes (metals basis)
Formula Weight 626.31
MDL Number MFCD00036272
UN Number UN3260
Merck Index 14,8777
Density 4.46g/mL
Packaging Ampule
Refractive Index 2.106
Sensitivity Moisture Sensitive
Boiling Point 340°C
Melting Point ≈143°C
Quantity 5g

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

  • Ampouled under argon