missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin(IV) iodide, 95%

Manufacturer:  Alfa Aesar™ 7111409

 View more versions of this product

Catalog No. AA7111409



Specifications

Specifications

Tin (IV) iodide
I4Sn
MFCD00036272
14,8777
Soluble in alcohol, benzene, CS2, chloroform, and ether; Decomposes in water
[Sn](I)(I)(I)I
626.328
626.31
95%
2.106
340°C
~143°C
7790-47-8
SnI4
UN3260
stannic iodide, stannane, tetraiodo, tin iv iodide, tetraiodotin, tin tetraiodide, tin iodide, unii-8h688bw69x, tin iv iodide 1:4, tin-iv-iodide, tin iv iodide
QPBYLOWPSRZOFX-UHFFFAOYSA-J
tetraiodostannane
24631
Crystalline
4.46g/mL
Moisture Sensitive
Red
10g

Chemical Identifiers

7790-47-8
626.328
QPBYLOWPSRZOFX-UHFFFAOYSA-J
24631
[Sn](I)(I)(I)I
I4Sn
MFCD00036272
stannic iodide, stannane, tetraiodo, tin iv iodide, tetraiodotin, tin tetraiodide, tin iodide, unii-8h688bw69x, tin iv iodide 1:4, tin-iv-iodide, tin iv iodide
tetraiodostannane
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

RTECSNumber : XQ3675000

TSCA : TSCA

SDS Documents