Learn More
Tin(IV) iodide, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 07111418
Description
Used in chemical research and as a food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
340°C | |
∼143°C | |
Red | |
50 g | |
SnI4 | |
UN3260 | |
stannic iodide, stannane, tetraiodo, tin iv iodide, tetraiodotin, tin tetraiodide, tin iodide, unii-8h688bw69x, tin iv iodide 1:4, tin-iv-iodide | |
ZBCVLXKKLMNLGV-UHFFFAOYSA-N | |
tetraiodostannane | |
24631 | |
95% | |
Moisture sensitive |
Tin (IV) iodide | |
7790-47-8 | |
Crystalline | |
H4I4Sn | |
MFCD00036272 | |
14,8777 | |
Soluble in alcohol,benzene,CS2,chloroform,and ether; Decomposes in water | |
[Sn].I.I.I.I | |
630.36 | |
626.31 | |
2.106 | |
4.46 g/mL |
Chemical Identifiers
7790-47-8 | |
630.36 | |
ZBCVLXKKLMNLGV-UHFFFAOYSA-N | |
24631 | |
[Sn].I.I.I.I |
H4I4Sn | |
MFCD00036272 | |
stannic iodide, stannane, tetraiodo, tin iv iodide, tetraiodotin, tin tetraiodide, tin iodide, unii-8h688bw69x, tin iv iodide 1:4, tin-iv-iodide | |
tetraiodostannane |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 232-208-4
RTECSNumber : XQ3675000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only