Learn More
Tin(IV) fluoride, 99% (metals basis), Thermo Scientific Chemicals
Friedel-Crafts catalyst
Supplier: Thermo Scientific Chemicals 01306304
Description
It is used as Friedel-Crafts catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tin(IV) fluoride | |
7783-62-2 | |
2 g | |
MFCD00042541 | |
14,8776 | |
99% |
705°C (sublimation) | |
Crystalline | |
(metals basis) | |
UN2923 | |
194.68 | |
Moisture sensitive |
Chemical Identifiers
7783-62-2 | |
MFCD00042541 |
Safety and Handling
GHS H Statement
H300-H314-H318
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber : 232-016-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only