Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin(IV) dibromide bis(2,4-pentanedionate), 98%

Chemical Identifiers

Chemical Name or Material Tin(IV) dibromide bis(2,4-pentanedionate)
CAS 16894-10-3
Formula Weight 476.72g/mol
MDL Number MFCD00075325
UN Number UN3261
Synonym Tin(IV) bis(acetylacetonate) dibromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5553203

 
alfa aesar™
H55532-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5553206

 
alfa aesar™
H55532-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5553214

 
alfa aesar™
H55532-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color White to yellow
Melting Point 182 to 184°C
Quantity 1g
Chemical Name or Material Tin(IV) dibromide bis(2,4-pentanedionate)
CAS 16894-10-3
Physical Form Powder
Assay 98%
Formula Weight 476.72g/mol
MDL Number MFCD00075325
UN Number UN3261
Synonym Tin(IV) bis(acetylacetonate) dibromide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container according to local regulations.
DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container according to local regulations.

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.