Learn More
Tin(IV) chloride hydrate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01157022
Description
Tin(IV) chloride hydrate is used in the formation of a xerogel via a cyanogel, a gel that has potential use in solid-state gas sensors. It is involved in the preparation of Indium tin oxide thin films by chemical vapor deposition method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tin(IV) chloride hydrate | |
7646-78-8 | |
100 g | |
Cl4Sn | |
tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4 | |
HPGGPRDJHPYFRM-UHFFFAOYSA-J | |
tetrachlorostannane | |
24287 |
56°C | |
Lump | |
98% | |
MFCD00149864 | |
Soluble in water and alcohol. | |
Cl[Sn](Cl)(Cl)Cl | |
260.51 | |
260.50 (Anhydrous) |
Chemical Identifiers
7646-78-8 | |
260.51 | |
HPGGPRDJHPYFRM-UHFFFAOYSA-J | |
24287 | |
Cl[Sn](Cl)(Cl)Cl |
Cl4Sn | |
MFCD00149864 | |
tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4 | |
tetrachlorostannane |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 231-588-9
RTECSNumber : XP8750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only