missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin(IV) chloride, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 7120314

 View more versions of this product

Catalog No. AA7120314



Specifications

Specifications

Tin(IV) chloride
99.99% (metals basis)
MFCD00011242
HPGGPRDJHPYFRM-UHFFFAOYSA-J
tetrachlorostannane
24287
Liquid
25g
7646-78-8
Cl4Sn
tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4
Cl[Sn](Cl)(Cl)Cl
260.51
260.5
2.26 g/mL

Chemical Identifiers

7646-78-8
260.51
HPGGPRDJHPYFRM-UHFFFAOYSA-J
24287
Cl[Sn](Cl)(Cl)Cl
Cl4Sn
MFCD00011242
tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4
tetrachlorostannane
Safety and Handling

Safety and Handling

EINECSNumber : 231-588-9

SDS Documents