Please login to your online account to display your discounted pricing

Tin(IV) chloride, 1M Solution in Dichloromethane, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC378201000

 View more versions of this product

Catalog No. AC378201000

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Tin(IV) chloride
Name Note 1M Solution in Dichloromethane
CAS 7646-78-8
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage. May cause respiratory irritation. Suspected of causing cancer. May cause drowsiness or dizziness.
CAS 75-09-2
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Molecular Formula Cl4Sn
Formula Weight 260.52
Appearance of Solution Clear colorless solution, fuming in air
MDL Number MFCD00011242
Fieser 01,1111; 03,269; 05,627; 06,553; 07,342; 09,436; 10,370; 11,552; 12,486; 13,300; 14,304; 17,335
Merck Index 15, 8901
Concentration 1M, exact strength on the certificate of analysis
Density 1.4190g/mL
Packaging AcroSeal™ Glass Bottle in Tin Can
Quantity 100mL

Specifications

Chemical Name or Material Tin(IV) chloride
Name Note 1M Solution in Dichloromethane
CAS 7646-78-8
CAS 75-09-2
Molecular Formula Cl4Sn
Formula Weight 260.52
MDL Number MFCD00011242
Accessories

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS H Statement
Causes severe skin burns and eye damage. May cause respiratory irritation. Suspected of causing cancer. May cause drowsiness or dizziness.

SDS & Safety Documents