Learn More
Tin(IV) acetate, 98%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 380050050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tin(IV) acetate | |
2800-96-6 | |
Crystalline Powder | |
98% | |
(CH3COO)4Sn | |
02, III, 134 | |
Solubility in water: hydrolysis. Other solubilities: hydroylsis with water to sn(iv) oxide hydrate and, acetic acid;, soluble in acetone, benzene;, less soluble in carbon tetrachloride | |
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sn+4] | |
354.89 | |
354.89 | |
Extra Pure | |
Glass bottle |
231.0°C to 237.0°C | |
White | |
5 g | |
C8H12O8Sn | |
MFCD00014976 | |
tin iv acetate, tin acetate, tetraacetoxystannane, tin-iv-acetate, acmc-20ajhv, tin 4+ ion tetraacetate, tetraacetoxytin, tin 4+ tetraacetate | |
YJGJRYWNNHUESM-UHFFFAOYSA-J | |
tin(4+);tetraacetate | |
9863446 | |
98% | |
Authentic |
Chemical Identifiers
2800-96-6 | |
354.89 | |
YJGJRYWNNHUESM-UHFFFAOYSA-J | |
9863446 | |
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sn+4] |
C8H12O8Sn | |
MFCD00014976 | |
tin iv acetate, tin acetate, tetraacetoxystannane, tin-iv-acetate, acmc-20ajhv, tin 4+ ion tetraacetate, tetraacetoxytin, tin 4+ tetraacetate | |
tin(4+);tetraacetate |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Risk of explosion if heated under confinement.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only