missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin(IV) acetate, 98%, ACROS Organics™

$191.29 - $191.29

Chemical Identifiers

CAS 2800-96-6
Assay Percent Range 98%
Molecular Formula C8H12O8Sn
Linear Formula (CH3COO)4Sn
Formula Weight 354.89
MDL Number MFCD00014976
Chemical Name or Material Tin(IV) acetate
Grade Extra Pure
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC380050010 SDS Thermo Fisher Scientific
AC380050010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC380050050 SDS Thermo Fisher Scientific
AC380050050
Glass bottle 5g Each for $191.29
Specifications Description & Specifications

Chemical Identifiers

2800-96-6
C8H12O8Sn
354.89
Tin(IV) acetate
98%
(CH3COO)4Sn
MFCD00014976
Extra Pure

Specifications

2800-96-6
98%
(CH3COO)4Sn
MFCD00014976
97.5% min.
Glass bottle
231° to 237°C
Tin(IV) acetate
Crystalline Powder
C8H12O8Sn
354.89
02, III, 134
Authentic
White
1g
Extra Pure

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Risk of explosion if heated under confinement.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning