Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin ingot/bar, 99.99% (metals basis), ASTM-B339-95b

Chemical Identifiers

Chemical Name or Material Tin ingot/bar
CAS 7440-31-5
Name Note ASTM-B339-95
MDL Number MFCD00133862
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA98394P2

 
alfa aesar™
98394P2
1 lb. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

MDL Number MFCD00133862
Name Note ASTM-B339-95
RTECS Number XP7320000
Assay 99.99%
Odor Odorless
Quantity 1 lb.
TSCA TSCA
Chemical Name or Material Tin ingot/bar
CAS 7440-31-5
Assay Percent Notes (metals basis)