Please login to your online account to display your discounted pricing

Tin(II) trifluoromethanesulfonate, 98%, ACROS Organics™


Quantity 5g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC312530050

 View more versions of this product

Catalog No. AC312530050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (ex Sn)
Physical Form Powder
Formula Weight 416.85
Quantity 5g
Color White to yellow
Chemical Name or Material Tin(II) trifluoromethanesulfonate
Health Hazard 1 GHS Signal Word: Danger
Assay Percent Range 98%
Packaging Plastic bottle
Infrared Spectrum Authentic
Molecular Formula C2F6O6S2Sn
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 62086-04-8
MDL Number MFCD00191251
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

Specifications

Chemical Name or Material Tin(II) trifluoromethanesulfonate
CAS 62086-04-8
Assay ≥97.5% (ex Sn)
Assay Percent Range 98%
Molecular Formula C2F6O6S2Sn
Formula Weight 416.85
MDL Number MFCD00191251
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents