missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin(II) telluride, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 8918618

 View more versions of this product

Catalog No. AA8918618



Specifications

Specifications

Tin (II) telluride
(metals basis)
MFCD00049543
246.29
99.999%
Odorless
780°C
12040-02-7
SnTe
UN3288
Pieces
6.48g/mL
3 to 12mm
50g

Chemical Identifiers

12040-02-7
SnTe
MFCD00049543
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents