missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin(II) sulfide, 99.5%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1405106

 View more versions of this product

Catalog No. AA1405106



Specifications

Specifications

Tin(II) sulfide
99.5%
MFCD00011245
AFNRRBXCCXDRPS-UHFFFAOYSA-N
sulfanylidenetin
426379
-8 Mesh
1314-95-0
SSn
tin ii sulfide, stannous sulfide, tin ii sulfide, tin sulfide sns, tin monosulfide, tin sulphide, stannanethione, herzenbergite, zinnsulfur, acmc-1btyn
S=[Sn]
150.77
150.75
5g

Chemical Identifiers

1314-95-0
150.77
AFNRRBXCCXDRPS-UHFFFAOYSA-N
426379
S=[Sn]
SSn
MFCD00011245
tin ii sulfide, stannous sulfide, tin ii sulfide, tin sulfide sns, tin monosulfide, tin sulphide, stannanethione, herzenbergite, zinnsulfur, acmc-1btyn
sulfanylidenetin
Safety and Handling

Safety and Handling

EINECSNumber : 215-248-7

Documents