Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin(II) stearate

Chemical Identifiers

Chemical Name or Material Tin(II) stearate
CAS 6994-59-8
Formula Weight 685.69
MDL Number MFCD00050720
UN Number UN3146
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3960522

  SDS
alfa aesar™
3960522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3960536

  SDS
alfa aesar™
3960536
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 685.69
Physical Form Powder/Chunks
Health Hazard 1 Warning
MDL Number MFCD00050720
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
Chemical Name or Material Tin(II) stearate
Quantity 100g
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 6994-59-8
UN Number UN3146