Learn More
Tin(II) pyrophosphate, Thermo Scientific Chemicals
$83.15 - $201.08
Chemical Identifiers
CAS | 15578-26-4 |
---|---|
Molecular Formula | O7P2Sn2 |
Molecular Weight (g/mol) | 411.361 |
MDL Number | MFCD00049544 |
InChI Key | GEZAUFNYMZVOFV-UHFFFAOYSA-J |
Synonym | stannous pyrophosphate, technescan pyp, ditin pyrophosphate, tin ii pyrophosphate, tin 2+ pyrophosphate, unii-4dnt29ec86, stannous pyrophosphate usan, tin 2+ diphosphate 1:2, pyro stannous phosphate, ditin 2+ pyrophosphate 4- |
PubChem CID | 66379 |
IUPAC Name | phosphonato phosphate;tin(2+) |
SMILES | [O-]P(=O)([O-])OP(=O)([O-])[O-].[Sn+2].[Sn+2] |
Description
Tin(II) pyrophosphate is used to inhibit the formation of volatile sulfur compounds. Further, it is used for cyanide-free tin coating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15578-26-4 | |
411.361 | |
GEZAUFNYMZVOFV-UHFFFAOYSA-J | |
66379 | |
[O-]P(=O)([O-])OP(=O)([O-])[O-].[Sn+2].[Sn+2] |
O7P2Sn2 | |
MFCD00049544 | |
stannous pyrophosphate, technescan pyp, ditin pyrophosphate, tin ii pyrophosphate, tin 2+ pyrophosphate, unii-4dnt29ec86, stannous pyrophosphate usan, tin 2+ diphosphate 1:2, pyro stannous phosphate, ditin 2+ pyrophosphate 4- | |
phosphonato phosphate;tin(2+) |
Specifications
400°C (decomposition) | |
Powder | |
O7P2Sn2 | |
stannous pyrophosphate, technescan pyp, ditin pyrophosphate, tin ii pyrophosphate, tin 2+ pyrophosphate, unii-4dnt29ec86, stannous pyrophosphate usan, tin 2+ diphosphate 1:2, pyro stannous phosphate, ditin 2+ pyrophosphate 4- | |
GEZAUFNYMZVOFV-UHFFFAOYSA-J | |
phosphonato phosphate;tin(2+) | |
66379 | |
4.009 |
15578-26-4 | |
50 g | |
MFCD00049544 | |
Insoluble in water. | |
[O-]P(=O)([O-])OP(=O)([O-])[O-].[Sn+2].[Sn+2] | |
411.361 | |
411.32 | |
Tin(II) Pyrophosphate |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
Irritating to eyes, respiratory system and skin.
EINECSNumber : 239-635-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only