missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin(II) oxide, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1156914

 View more versions of this product

Catalog No. AA1156914



Specifications

Specifications

Tin(II) oxide
99.9% (metals basis)
MFCD00011243
QHGNHLZPVBIIPX-UHFFFAOYSA-N
oxotin
88989
-100 Mesh Powder
25g
21651-19-4
OSn
oxo stannane
O=[Sn]
134.709
134.69
6.45g/mL

Chemical Identifiers

21651-19-4
134.709
QHGNHLZPVBIIPX-UHFFFAOYSA-N
88989
O=[Sn]
OSn
MFCD00011243
oxo stannane
oxotin
Safety and Handling

Safety and Handling

EINECSNumber : 244-499-5

Documents