Please login to your online account to display your discounted pricing

Tin(II) oxide, 99.9% (metals basis), Alfa Aesar™

Reducing agent, soft abrasive, preparation of stannous salts

Chemical Identifiers

Chemical Name or Material Tin(II) oxide
CAS 21651-19-4
Molecular Formula OSn
Linear Formula SnO
Formula Weight 134.69
MDL Number MFCD00011243
Merck Index 14,8787
Synonym Stannous oxide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1156936

  SDS
alfa aesar
1156936
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 134.69
Quantity 500g
Chemical Name or Material Tin(II) oxide
TSCA TSCA
Mesh Size -100 Mesh
Odor Odorless
Physical Form Powder
Assay 99.9%
Assay Percent Notes (metals basis)
Molecular Formula OSn
Linear Formula SnO
MDL Number MFCD00011243
CAS 21651-19-4
Merck Index 14,8787
Synonym Stannous oxide
Color Blue-black
Melting Point 1080°C (dec.)
Solubility Information Insoluble in water, alcohol. Soluble in acids, and concentrated NaOH or KOH solutions
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302;
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a;
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 6.45
RTECS Number XQ3700000
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a;
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302;
Harmful if swallowed.