missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin(II) oxalate

Manufacturer:  Alfa Aesar™ 1411322

 View more versions of this product

Catalog No. AA1411322



Specifications

Specifications

Tin(II) oxalate
814-94-8
3.56g/mL
C2O4Sn
MFCD00040678
3708588
stannous oxalate, tin ii oxalate, tin oxalate, tin 2+ oxalate, ethanedioic acid, tin 2+ salt 1:1, stavelan cinaty czech, unii-sar72fe8eh, tin ii oxalate, sar72fe8eh, stavelan cinaty
OQBLGYCUQGDOOR-UHFFFAOYSA-L
oxalate;tin(2+)
13149
Powder
280°C (decomposition)
100g
White
SnC2O4
UN3288
14,8786
Soluble in dilute HCl; Insoluble in water
C(=O)(C(=O)[O-])[O-].[Sn+2]
206.728
206.72

Chemical Identifiers

814-94-8
206.728
OQBLGYCUQGDOOR-UHFFFAOYSA-L
13149
C(=O)(C(=O)[O-])[O-].[Sn+2]
C2O4Sn
MFCD00040678
stannous oxalate, tin ii oxalate, tin oxalate, tin 2+ oxalate, ethanedioic acid, tin 2+ salt 1:1, stavelan cinaty czech, unii-sar72fe8eh, tin ii oxalate, sar72fe8eh, stavelan cinaty
oxalate;tin(2+)
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

RTECSNumber : XQ3950000

TSCA : TSCA

Documents