Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin(II) methoxide

$76.05 - $76.05

Chemical Identifiers

Chemical Name or Material Tin(II) methoxide
CAS 14794-99-1
Formula Weight 180.76
MDL Number MFCD00015612
Synonym Tetramethoxytin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2089706

  SDS
alfa aesar™
2089706
5g Each for $76.05

AA2089714

  SDS
alfa aesar™
2089714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.76
Chemical Name or Material Tin(II) methoxide
Melting Point 242° to 243°C
Physical Form Powder
MDL Number MFCD00015612
CAS 14794-99-1
Quantity 5g
Synonym Tetramethoxytin
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Moisture Sensitive
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.