missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin(II) Iodide, 99.99% (Metal Basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1447614

 View more versions of this product

Catalog No. AA1447614



Specifications

Specifications

Tin(II) Iodide
10294-70-9
I2Sn
tin ii iodide, stannous iodide, tin diiodide, tin iodide sni2, stannous diiodide, sni2, acmc-1bqot, tin ii iodide, ultra dry, tin ii iodide,-10 mesh trace metals basis, tin ii iodide, anhydrous, beads,-10 mesh trace metals basis
[Sn](I)I
372.519
372.5
Air, moisture sensitive
Ampouled under argon
99.99%, metal basis
MFCD00049545
JTDNNCYXCFHBGG-UHFFFAOYSA-L
diiodotin
25138
-10 Mesh Beads
25g

Chemical Identifiers

10294-70-9
372.519
JTDNNCYXCFHBGG-UHFFFAOYSA-L
25138
[Sn](I)I
I2Sn
MFCD00049545
tin ii iodide, stannous iodide, tin diiodide, tin iodide sni2, stannous diiodide, sni2, acmc-1bqot, tin ii iodide, ultra dry, tin ii iodide,-10 mesh trace metals basis, tin ii iodide, anhydrous, beads,-10 mesh trace metals basis
diiodotin
Safety and Handling

Safety and Handling

When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

Causes burns.

DOTInformation : DOT Class: 8

EINECSNumber : 233-667-3

SDS Documents