missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tin(II) fluorophosphate, Thermo Scientific Chemicals
$106.64 - $106.64
Chemical Identifiers
CAS | 52262-58-5 |
---|---|
Molecular Formula | FO3PSn |
Molecular Weight (g/mol) | 216.679 |
MDL Number | MFCD00058794 |
InChI Key | ZIQLCYCKKROENE-UHFFFAOYSA-L |
Synonym | tin ii fluorophosphate, stannous fluorophosphate, acmc-20ajhz, fluoridophosphoric acid tin ii salt, tin, ion sn2+ fluoro-phosphite ion, fluoro-dioxido-oxophosphorane; tin 2+, fluoranyl-bis oxidanidyl-oxidanylidene-$l^ 5-phosphane; tin 2+ |
PubChem CID | 44717639 |
IUPAC Name | fluoro-dioxido-oxo-$l^{5}-phosphane;tin(2+) |
SMILES | [O-]P(=O)([O-])F.[Sn+2] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA3960806
|
Thermo Scientific Chemicals
03960806 |
5 g |
Each for $106.64
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
52262-58-5 | |
216.679 | |
ZIQLCYCKKROENE-UHFFFAOYSA-L | |
44717639 | |
[O-]P(=O)([O-])F.[Sn+2] |
FO3PSn | |
MFCD00058794 | |
tin ii fluorophosphate, stannous fluorophosphate, acmc-20ajhz, fluoridophosphoric acid tin ii salt, tin, ion sn2+ fluoro-phosphite ion, fluoro-dioxido-oxophosphorane; tin 2+, fluoranyl-bis oxidanidyl-oxidanylidene-$l^ 5-phosphane; tin 2+ | |
fluoro-dioxido-oxo-$l^{5}-phosphane;tin(2+) |
Specifications
White | |
Powder | |
FO3PSn | |
tin ii fluorophosphate, stannous fluorophosphate, acmc-20ajhz, fluoridophosphoric acid tin ii salt, tin, ion sn2+ fluoro-phosphite ion, fluoro-dioxido-oxophosphorane; tin 2+, fluoranyl-bis oxidanidyl-oxidanylidene-$l^ 5-phosphane; tin 2+ | |
ZIQLCYCKKROENE-UHFFFAOYSA-L | |
fluoro-dioxido-oxo-$l^{5}-phosphane;tin(2+) | |
44717639 | |
Tin(II) fluorophosphate |
52262-58-5 | |
5 g | |
MFCD00058794 | |
Slightly soluble (3.0g/L at 25°C) | |
[O-]P(=O)([O-])F.[Sn+2] | |
216.679 | |
216.66 |
Safety and Handling
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only