missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tin(II) ethoxide

Manufacturer:  Alfa Aesar™ 2089606

 View more versions of this product

Catalog No. AA2089606



Specifications

Specifications

Tin(II) ethoxide
14791-99-2
Air & Moisture Sensitive
Sn(OC2H5)2
Reacts with water
CC[O-].CC[O-].[Sn+2]
208.832
208.81
~200°C (decomposition)
5g
C4H10O2Sn
MFCD00015653
XCKWFNSALCEAPW-UHFFFAOYSA-N
ethanolate;tin(2+)
71317418
Solid/Semi-Solid

Chemical Identifiers

14791-99-2
208.832
XCKWFNSALCEAPW-UHFFFAOYSA-N
ethanolate;tin(2+)
C4H10O2Sn
MFCD00015653
71317418
CC[O-].CC[O-].[Sn+2]
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents